4-Bromo-3-hydroxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30823-1G 30823-25G 30823-500MG 30823-5G
76930-432EA 350.1 USD
76930-432 76930-434 76930-436 76930-438
4-Bromo-3-hydroxyphenylboronic acid pinacol ester ≥97%
4-Bromo-3-hydroxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BBrO₃
MW: 298.969 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16999888
CAS Number: 2121512-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR