4-Chloro-3-hydroxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Chloro-3-hydroxyphenyl)boronic acid

30806-1G 30806-25G 30806-500MG 30806-5G
76930-416EA 188.85 USD
76930-416 76930-418 76930-420 76930-422
4-Chloro-3-hydroxyphenylboronic acid ≥97%
4-Chloro-3-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BClO₃
MW: 172.3740 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10697413
CAS Number: 915201-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR