4-Fluoro-2-hydroxybenzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 4-Fluoro-2-hydroxyphenylboronic acid pinacol ester

30782-1G 30782-25G 30782-500MG 30782-5G
76930-386EA 260.99 USD
76930-386 76930-388 76930-390 76930-392
4-Fluoro-2-hydroxybenzeneboronic acid pinacol ester ≥97%
4-Fluoro-2-hydroxybenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BFO₃
MW: 238.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16994248
CAS Number: 1038828-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR