5-Bromo-3-iodo-2,6-dimethoxypyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
30733-25G
30733-5G
76930-364EA
453.01
USD
76930-364
76930-366
5-Bromo-3-iodo-2,6-dimethoxypyridine ≥97%
5-Bromo-3-iodo-2,6-dimethoxypyridine
Precaución: for Research Use Only
Formula:
C₇H₇BrINO₂ MW: 343.944 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29088182 CAS Number: 1826110-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...