4-Bromo-2-fluoro-6-iodophenol ≥97%

Supplier: AOBChem USA

Warning

30452-1G 30452-25G 30452-500MG 30452-5G
76930-308EA 138.98 USD
76930-308 76930-310 76930-312 76930-314
4-Bromo-2-fluoro-6-iodophenol ≥97%
4-Bromo-2-fluoro-6-iodophenol

Precaución: for Research Use Only

Formula: C₆H₃BrFIO
Storage Temperature: Refrigerator
MDL Number: MFCD26895840
CAS Number: 1228093-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR