5-Chloro-2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 5-Chloro-2-fluoropyridine-3-boronic acid pinacol ester

30388-25G 30388-5G
76930-272EA 595.17 USD
76930-272 76930-274
5-Chloro-2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97% research grade
5-Chloro-2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Formula: C₁₁H₁₄BClFNO₂
MW: 257.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08063076
CAS Number: 937595-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR