4-Chloro-2,3-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

30357-1G 30357-25G 30357-500MG 30357-5G
76930-256EA 379.81 USD
76930-256 76930-258 76930-260 76930-262
4-Chloro-2,3-dimethylbenzaldehyde ≥97%
4-Chloro-2,3-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO
MW: 168.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917146
CAS Number: 1349716-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR