5-Chloro-2-isopropoxy-3-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (5-Chloro-2-isopropoxypyridin-3-yl)boronic acid, 5-Chloro-2-isopropoxypyridine-3-boronic acid

30330-1G 30330-25G 30330-5G
76930-244EA 102.91 USD
76930-244 76930-246 76930-248
5-Chloro-2-isopropoxy-3-pyridineboronic acid ≥97%
5-Chloro-2-isopropoxy-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BClNO₃
Storage Temperature: Ambient
MDL Number: MFCD13195662
CAS Number: 1217501-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR