5-Chloro-2-ethoxy-pyridine-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

30326-1G 30326-25G 30326-500MG 30326-5G
76930-232EA 379.81 USD
76930-232 76930-234 76930-236 76930-238
5-Chloro-2-ethoxy-pyridine-3-carbaldehyde ≥97%
5-Chloro-2-ethoxy-pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD18254513
CAS Number: 870721-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR