5-Bromo-2-(bromomethyl)pyridine hydrobromide ≥95%
Supplier: AOBChem USA
|
Warning
|
30243-1G
30243-25G
30243-5G
76930-218EA
90.18
USD
76930-218
76930-220
76930-222
5-Bromo-2-(bromomethyl)pyridine hydrobromide ≥95%
5-Bromo-2-(bromomethyl)pyridine hydrobromide
Formula:
C₆H₆Br₃N MW: 331.83 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20921831 CAS Number: 173999-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...