5-Bromo-2-(propan-2-yloxy)pyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

30236-1G 30236-25G 30236-500MG 30236-5G
76930-202EA 254.62 USD
76930-202 76930-204 76930-206 76930-208
5-Bromo-2-(propan-2-yloxy)pyridin-3-ol ≥97%
5-Bromo-2-(propan-2-yloxy)pyridin-3-ol

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO₂
MW: 232.075 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29088148
CAS Number: 1826110-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR