4-Bromo-2-methylbiphenyl ≥97%

Supplier: AOBChem USA

Warning

30191-25G 30191-5G
76930-190EA 595.17 USD
76930-190 76930-192
4-Bromo-2-methylbiphenyl ≥97%
4-Bromo-2-methylbiphenyl
Formula: C₁₃H₁₁Br
Storage Temperature: Ambient
MDL Number: MFCD03093069
CAS Number: 5002-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR