4-Bromo-2-fluoro-3-hydroxyanisole ≥97%

Supplier: AOBChem USA

Warning

30152-1G 30152-25G 30152-5G
76930-166EA 106.09 USD
76930-166 76930-168 76930-170
4-Bromo-2-fluoro-3-hydroxyanisole ≥97%
4-Bromo-2-fluoro-3-hydroxyanisole

Precaución: for Research Use Only

Formula: C₇H₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28738847
CAS Number: 1784923-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR