2-Ethoxy-5-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Fluoro-2-(ethoxy)-pyridine-4-boronic acid pinacol ester

30063-1G 30063-25G 30063-5G
76930-152EA 96.55 USD
76930-152 76930-154 76930-156
2-Ethoxy-5-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%
2-Ethoxy-5-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Formula: C₁₃H₁₉BFNO₃
MW: 267.1043 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28054670
CAS Number: 2121513-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR