5-Bromo-2,3-difluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

29671-1G 29671-25G 29671-5G
76930-072EA 102.91 USD
76930-072 76930-074 76930-076
5-Bromo-2,3-difluorobenzoic acid ≥97%
5-Bromo-2,3-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04972472
CAS Number: 887585-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR