4-Isopropoxy-3-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

29572-25G 29572-5G
76930-062EA 453.01 USD
76930-062 76930-064
4-Isopropoxy-3-methoxybenzoic acid ≥97%
4-Isopropoxy-3-methoxybenzoic acid
Formula: C₁₁H₁₄O₄
Storage Temperature: Refrigerator
MDL Number: MFCD01993664
CAS Number: 3535-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR