4'-Fluoro-3'-methyl-[1,1'-biphenyl]-4-ol ≥97%

Supplier: AOBChem USA

Warning

29563-1G 29563-25G 29563-500MG 29563-5G
76930-050EA 189.91 USD
76930-050 76930-052 76930-054 76930-056
4'-Fluoro-3'-methyl-[1,1'-biphenyl]-4-ol ≥97%
4'-Fluoro-3'-methyl-[1,1'-biphenyl]-4-ol

Precaución: for Research Use Only

Formula: C₁₃H₁₁FO
MW: 202.224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18312810
CAS Number: 64465-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR