5-Bromo-2-(difluoromethyl)pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
29412-25G
29412-5G
76930-034EA
573.95
USD
76930-034
76930-036
5-Bromo-2-(difluoromethyl)pyridine ≥97%
5-Bromo-2-(difluoromethyl)pyridine
Formula:
C₆H₄BrF₂N Storage Temperature: Ambient |
MDL Number:
MFCD11977429 CAS Number: 845827-13-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...