4-Isopropyl-3-nitroaniline ≥97%

Supplier: AOBChem USA

Warning

29331-1G 29331-25G 29331-500MG 29331-5G
76930-006EA 260.99 USD
76930-006 76930-008 76930-010 76930-012
4-Isopropyl-3-nitroaniline ≥97%
4-Isopropyl-3-nitroaniline

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00092573
CAS Number: 92765-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR