4-Cyclopropyl-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

29284-1G 29284-25G 29284-500MG 29284-5G
76929-992EA 228.1 USD
76929-992 76929-994 76929-996 76929-998
4-Cyclopropyl-2-methylbenzoic acid ≥97%
4-Cyclopropyl-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD20645932
CAS Number: 909698-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR