4-Methoxy-2-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Methoxy-2-trifluormethylbenzoic acid

29210-25G 29210-5G
76929-980EA 315.09 USD
76929-980 76929-982
4-Methoxy-2-(trifluoromethyl)benzoic acid ≥97%
4-Methoxy-2-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01091013
CAS Number: 127817-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR