4-Bromo-3-ethylphenol ≥97% research grade

Supplier: AOBChem USA

Warning

29167-1G 29167-25G 29167-500MG 29167-5G
76929-966EA 168.69 USD
76929-966 76929-968 76929-970 76929-972
4-Bromo-3-ethylphenol ≥97% research grade
4-Bromo-3-ethylphenol
Formula: C₈H₉BrO
Storage Temperature: Refrigerator
MDL Number: MFCD16997652
CAS Number: 99873-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR