5-Bromo-2-(3-fluorophenoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

28776-1G 28776-25G 28776-500MG 28776-5G
76929-896EA 202.64 USD
76929-896 76929-898 76929-900 76929-902
5-Bromo-2-(3-fluorophenoxy)pyridine ≥97%
5-Bromo-2-(3-fluorophenoxy)pyridine
Formula: C₁₁H₇BrFNO
Storage Temperature: Ambient
MDL Number: MFCD14652062
CAS Number: 936343-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR