5-(Trifluoromethyl)pyridine-2-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-(Trifluoromethyl)picolinaldehyde

28687-25G 28687-5G
76929-878EA 519.85 USD
76929-878 76929-880
5-(Trifluoromethyl)pyridine-2-carboxaldehyde ≥97%
5-(Trifluoromethyl)pyridine-2-carboxaldehyde
Formula: C₇H₄F₃NO
MW: 175.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07375388
CAS Number: 31224-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR