4-Cyano-3-ethylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

28569-1G 28569-500MG 28569-5G
76929-840EA 795.68 USD
76929-840 76929-842 76929-844
4-Cyano-3-ethylphenylboronic acid ≥95%
4-Cyano-3-ethylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BNO₂
MW: 174.992 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22491274
CAS Number: 911210-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR