4-Hydroxy-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

28527-1G 28527-25G 28527-500MG 28527-5G
76929-824EA 231.28 USD
76929-824 76929-826 76929-828 76929-830
4-Hydroxy-2-methoxybenzoic acid ≥97%
4-Hydroxy-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈O₄
Storage Temperature: Refrigerator
MDL Number: MFCD06203226
CAS Number: 90111-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR