5-Bromo-2-(1H-imidazol-1-yl)pyridine ≥97%

Supplier: AOBChem USA

Warning

28479-1G 28479-25G 28479-5G
76929-798EA 109.28 USD
76929-798 76929-800 76929-802
5-Bromo-2-(1H-imidazol-1-yl)pyridine ≥97%
5-Bromo-2-(1H-imidazol-1-yl)pyridine

Precaución: for Research Use Only

Formula: C₈H₆BrN₃
MW: 224.057 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08668594
CAS Number: 380381-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR