5-Ethylthiophene-2-carboxylic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 5-Ethyl-thiophene-2-carboxylic acid

28347-1G 28347-25G 28347-5G
76929-772EA 62.6 USD
76929-772 76929-774 76929-776
5-Ethylthiophene-2-carboxylic acid ≥97% research grade
5-Ethylthiophene-2-carboxylic acid
Formula: C₇H₈O₂S
Storage Temperature: Ambient
MDL Number: MFCD00130089
CAS Number: 23229-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR