5-Hexyl-2-thiophenecarboxylic acid ≥97%

Supplier: AOBChem USA

Warning

28325-1G 28325-25G 28325-5G
76929-760EA 62.6 USD
76929-760 76929-762 76929-764
5-Hexyl-2-thiophenecarboxylic acid ≥97%
5-Hexyl-2-thiophenecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD01927415
CAS Number: 90619-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR