5-Bromo-3-(trifluoromethyl)salicylaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-hydroxy-3-(trifluoromethyl)benzaldehyde

28197-1G 28197-25G 28197-500MG 28197-5G
76929-712EA 144.29 USD
76929-712 76929-714 76929-716 76929-718
5-Bromo-3-(trifluoromethyl)salicylaldehyde ≥97%
5-Bromo-3-(trifluoromethyl)salicylaldehyde

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₂
MW: 269.200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03788531
CAS Number: 251300-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR