4-Fluorobenzofuran ≥97%

Supplier: AOBChem USA

Warning

28184-1G 28184-500MG 28184-5G
76929-700EA 797.8 USD
76929-700 76929-702 76929-704
4-Fluorobenzofuran ≥97%
4-Fluorobenzofuran
Formula: C₈H₅FO
MW: 136.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10699416
CAS Number: 24410-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR