5-Bromo-4-methylnicotinonitrile ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-4-methylpyridine-3-carbonitrile

28010-1G 28010-25G 28010-500MG 28010-5G
76929-680EA 254.62 USD
76929-680 76929-682 76929-684 76929-686
5-Bromo-4-methylnicotinonitrile ≥97%
5-Bromo-4-methylnicotinonitrile
Formula: C₇H₅BrN₂
MW: 197.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07439978
CAS Number: 890092-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR