5-Fluororesorcinol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Fluorobenzene-1,3-diol

27967-1G 27967-25G 27967-5G
76929-662EA 72.15 USD
76929-662 76929-664 76929-666
5-Fluororesorcinol ≥97%
5-Fluororesorcinol

Precaución: for Research Use Only

Formula: C₆H₅FO₂
MW: 128.10 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08457182
CAS Number: 75996-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR