4-Fluoronaphthalen-1-ol ≥97%

Supplier: AOBChem USA

Warning

27926-25G 27926-5G
76929-644EA 502.87 USD
76929-644 76929-646
4-Fluoronaphthalen-1-ol ≥97%
4-Fluoronaphthalen-1-ol
Formula: C₁₀H₇FO
Storage Temperature: Ambient
MDL Number: MFCD10699695
CAS Number: 315-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR