2-(4-Chloro-3-(trifluoromethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-3-(trifluoromethoxy)phenylboronic acid pinacol ester

27806-1G 27806-500MG 27806-5G
76929-632EA 468.92 USD
76929-632 76929-634 76929-636
2-(4-Chloro-3-(trifluoromethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-Chloro-3-(trifluoromethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₃H₁₅BClF₃O₃
MW: 322.5156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18756754
CAS Number: 2098632-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR