4-Phenoxy-2-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

27716-1G 27716-500MG 27716-5G
76929-616EA 1178.66 USD
76929-616 76929-618 76929-620
4-Phenoxy-2-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%
4-Phenoxy-2-(trifluoromethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₀BF₃O₃
MW: 364.167 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18384032
CAS Number: 1196396-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR