5'-Fluoro-2'-methylacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Fluoro-2-methylacetophenone, 1-(5-Fluoro-2-methylphenyl)ethan-1-one

27663-25G 27663-5G
76929-602EA 396.78 USD
76929-602 76929-604
5'-Fluoro-2'-methylacetophenone ≥97%
5'-Fluoro-2'-methylacetophenone

Precaución: for Research Use Only

Formula: C₉H₉FO
MW: 152.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06660189
CAS Number: 29427-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR