5-Ethoxy-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

27597-1G 27597-25G 27597-500MG 27597-5G
76929-578EA 242.95 USD
76929-578 76929-580 76929-582 76929-584
5-Ethoxy-2-methylphenylboronic acid ≥97%
5-Ethoxy-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₃
MW: 180.009 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28009350
CAS Number: 1383576-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR