4-Chloro-2-fluoro-5-methylbenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
27577-1G
27577-25G
27577-500MG
27577-5G
76929-562EA
290.69
USD
76929-562
76929-564
76929-566
76929-568
4-Chloro-2-fluoro-5-methylbenzoic acid ≥97%
4-Chloro-2-fluoro-5-methylbenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₆ClFO₂ MW: 188.583 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD20660427 CAS Number: 177211-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...