4-Bromo-2-fluoro-3-iodobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

27538-1G 27538-25G 27538-5G
76929-548EA 106.09 USD
76929-548 76929-550 76929-552
4-Bromo-2-fluoro-3-iodobenzoic acid ≥95%
4-Bromo-2-fluoro-3-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrFIO₂
MW: 344.904 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28142601
CAS Number: 1809157-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR