4-Bromo-3-chloro-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

27505-1G 27505-25G 27505-500MG 27505-5G
76929-518EA 194.15 USD
76929-518 76929-520 76929-522 76929-524
4-Bromo-3-chloro-2-fluorophenylboronic acid ≥97%
4-Bromo-3-chloro-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BBrClFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD28142599
CAS Number: 2121514-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR