4-Bromo-2-methylthioanisole ≥97%

Supplier: AOBChem USA

Warning

27382-1G 27382-25G 27382-5G
76929-480EA 62.6 USD
76929-480 76929-482 76929-484
4-Bromo-2-methylthioanisole ≥97%
4-Bromo-2-methylthioanisole

Precaución: for Research Use Only

Formula: C₈H₉BrS
MW: 217.126 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27931825
CAS Number: 89981-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR