5-Fluoro-3-pyrrolidin-1-ylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

27298-1G 27298-25G 27298-500MG 27298-5G
76929-464EA 195.21 USD
76929-464 76929-466 76929-468 76929-470
5-Fluoro-3-pyrrolidin-1-ylbenzaldehyde ≥95%
5-Fluoro-3-pyrrolidin-1-ylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₂FNO
MW: 193.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28015447
CAS Number: 1692606-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR