4-Bromo-3-iodoanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-2-iodo-4-methoxybenzene

27118-1G 27118-25G 27118-5G
76929-430EA 62.6 USD
76929-430 76929-432 76929-434
4-Bromo-3-iodoanisole ≥97%
4-Bromo-3-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₆BrIO
Storage Temperature: Refrigerator
MDL Number: MFCD00070743
CAS Number: 4897-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR