5-Bromo-4-methyl-2-(pyrrolidin-1-ylcarbonyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

27059-1G 27059-25G 27059-500MG 27059-5G
76929-406EA 194.15 USD
76929-406 76929-408 76929-410 76929-412
5-Bromo-4-methyl-2-(pyrrolidin-1-ylcarbonyl)pyridine ≥97%
5-Bromo-4-methyl-2-(pyrrolidin-1-ylcarbonyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrN₂O
MW: 269.138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28015436
CAS Number: 1809161-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR