5-Bromo-1-[(4-Methylphenyl)Sulfonyl]-1H-Indole ≥95%

Supplier: AOBChem USA

Warning

26973-25G 26973-5G
76929-360EA 453.01 USD
76929-360 76929-362
5-Bromo-1-[(4-Methylphenyl)Sulfonyl]-1H-Indole ≥95%
5-Bromo-1-[(4-Methylphenyl)Sulfonyl]-1H-Indole

Precaución: for Research Use Only

Formula: C₁₅H₁₂BrNO₂S
MW: 350.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20486680
CAS Number: 96546-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR