4-Hydroxy-2-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Hydroxy-2-(trifluoromethyl)phenylboronic acid, pinacol ester

26648-1G 26648-25G 26648-500MG 26648-5G
76929-278EA 374.5 USD
76929-278 76929-280 76929-282 76929-284
4-Hydroxy-2-(trifluoromethyl)phenylboronic acid pinacol ester ≥97%
4-Hydroxy-2-(trifluoromethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BF₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD15143616
CAS Number: 1029439-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR