5-Bromo-2-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

26550-1G 26550-25G 26550-5G
76929-256EA 117.76 USD
76929-256 76929-258 76929-260
5-Bromo-2-methylphenylboronic acid pinacol ester ≥97%
5-Bromo-2-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BBrO₂
MW: 296.996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12923176
CAS Number: 1192051-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR