4-Fluoro-3-formylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

26534-1G 26534-25G 26534-5G
76929-244EA 76.39 USD
76929-244 76929-246 76929-248
4-Fluoro-3-formylphenylboronic acid pinacol ester ≥97%
4-Fluoro-3-formylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BFO₃
MW: 250.0738 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846346
CAS Number: 443776-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR