4-Trifluoromethyl thioanisole ≥97%

Supplier: AOBChem USA

Warning

26532-1G 26532-25G 26532-500MG 26532-5G
76929-228EA 142.17 USD
76929-228 76929-230 76929-232 76929-234
4-Trifluoromethyl thioanisole ≥97%
4-Trifluoromethyl thioanisole

Precaución: for Research Use Only

Formula: C₈H₇F₃S
MW: 192.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09038792
CAS Number: 329-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR